Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dibutyl phthalate Monoclonal Antibody (4G3)

Catalog No. PIMA549627
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549627 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549627 Supplier Invitrogen™ Supplier No. MA549627

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Dibutyl phthalate (DBP) is a commonly used plasticizer that is added to various consumer products to increase their flexibility and durability. It is a clear, colorless liquid with a slightly sweet odor and is primarily used in the production of plastics, especially PVC. DBP is also found in adhesives, printing inks, sealants, and personal care products such as nail polish and hair sprays. It is known to be toxic to aquatic life and may also have adverse effects on human health, particularly on the reproductive system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dibutyl phthalate
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 4G3
Concentration 3.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias Dibutyl phthalate
Host Species Mouse
Immunogen Conjugate of DBP hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.