Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyl Succinate 99.0+%, TCI America™
SDP

Catalog No. S0102500ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 141-03-7
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.304
MDL Number MFCD00040495
InChI Key YUXIBTJKHLUKBD-UHFFFAOYSA-N
Synonym dibutyl succinate, tabutrex, succinic acid di-n-butyl ester, di-n-butyl succinate, tabatrex, dibutylsuccinate, butyl butanedioate, butanedioic acid, dibutyl ester, succinic acid dibutyl ester, dnbs
PubChem CID 8830
ChEBI CHEBI:82263
IUPAC Name dibutyl butanedioate
SMILES CCCCOC(=O)CCC(=O)OCCCC

Specifications

Melting Point -29°C
Color Colorless
Boiling Point 274°C
Quantity 500 mL
Formula Weight 230.30
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyl Succinate
EINECSNumber (2)-0848
RTECSNumber WM6250000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.