Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Dibutyltin dichloride, 96%
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                25 g
                            
                        
                            
                                250 g
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Glass bottle
                            
                        
                    Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
            Specifications
Specifications
| Melting Point | 39.0°C to 41.0°C | 
| Color | White to Beige | 
| Density | 1.4000g/mL | 
| Boiling Point | 135.0°C (10.0 mmHg) | 
| Flash Point | 112°C | 
| Infrared Spectrum | Authentic | 
| Assay Percent Range | 96% min | 
| Packaging | Glass bottle | 
| Molecular Formula | C8H18Cl2Sn | 
| Linear Formula | [CH3(CH2)3]2SnCl2 | 
| Show More | 
Safety and Handling
| Hazard Category | GHS Signal Word: Danger | 
| Hazard Statement | GHS H Statement Very toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. Suspected of causing genetic defects. Causes damage to organs through prolonged or repeated exposure. Harmful in contact with skin. Fatal if inhaled. Toxic if swallowed. May damage fertility. May damage the unborn child. | 
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. | 
| EINECSNumber | 211-670- | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                