Learn More
Description
For more information about how Dibutyryl-cAMP, sodium salt may be used, see our protocol: Generating Midbrain Dopaminergic Neurons from hPSCs
This product is Bio-Techne's ancillary Material Grade version of Dibutyryl-cAMP sodium salt (Cat. No. 1141). Suitable for use in research and further manufacturing of cell therapies. Find out more about our Ancillary Material Grade product range.
Specifications
Specifications
| Chemical Name or Material | N6,O2'-Dibutyryladenosine 3',5'-cyclic monophosphate sodium salt |
| CAS | 16980-89-5 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | Ancillary Material Grade Compounds |
| Molecular Formula | C18H23N5NaO8P |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
