Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dibutyryl-cAMP, sodium salt
GREENER_CHOICESDP

Catalog No. 114110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-411-0 10 mg
11-415-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-411-0 Supplier Tocris Bioscience™ Supplier No. 1141/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cell-permeable cAMP analog

Cell-permeable analog of cAMP; activates cAMP-dependent protein kinases. Promotes differentiation of dopaminergic neurons (iDA) from hPSCs in combination with other reagents.

Chemical Identifiers

CAS 16980-89-5
Molecular Formula C18H26N5NaO8P

Specifications

Quantity 10 mg
Formula Weight 491.37
Purity >98%
Molecular Weight (g/mol) 494.397
Chemical Name or Material Dibutyryl-cAMP, sodium salt
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.