Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicarbonylacetylacetonato rhodium(I)

Catalog No. AC363950025
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle

CAS: 14874-82-9 | C7H7O4Rh | 258.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14874-82-9
Molecular Formula C7H7O4Rh
Molecular Weight (g/mol) 258.03
MDL Number MFCD00009884
InChI Key BZCAWKOPWNIDOC-FGSKAQBVSA-M
IUPAC Name λ¹-rhodium(1+) bis(methanidylidyneoxidanium) (2Z)-4-oxopent-2-en-2-olate
SMILES [Rh+].[C-]#[O+].[C-]#[O+].C\C([O-])=C\C(C)=O

Specifications

Melting Point 154.0°C to 156.0°C
Color Green-Red
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 2.5 g
Solubility Information Solubility in water: insoluble
Formula Weight 258.04
Physical Form Flakes or Needles
Chemical Name or Material Dicarbonylacetylacetonato rhodium(I)
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Toxic if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Risk of explosion if heated under confinement
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occ
EINECSNumber 238-947-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.