Learn More
Invitrogen™ Dicer Monoclonal Antibody (GT1176)

Description
Positive Control: A431 Predicted Reactivity: Mouse (97%), Zebrafish (93%), Xenopus laevis (92%), Pig (99%), Chicken (96%), Rhesus Monkey (100%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Dicer |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT1176 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | DICER1 |
| Gene Accession No. | Q8R418, Q9UPY3 |
| Gene Alias | 1110006F08Rik; D12Ertd7e; DCR; DCR1; DCR-1; Dicer; dicer 1 ribonuclease III; dicer 1, double-stranded RNA-specific endoribonuclease; dicer 1, ribonuclease III; dicer 1, ribonuclease type III; Dicer1; Dicer1, Dcr-1 homolog; Dicer1e; double-strand-specific ribonuclease mDCR-1; endoribonuclease Dicer; Helicase MOI; helicase with RNAse motif; helicase-moi; HERNA; K12H4.8-LIKE; KIAA0928; mKIAA0928; MNG1; RMSE2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.