Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicer Monoclonal Antibody (S167-7), QED Bioscience™
SDP

Catalog No. 502277468
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7468 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7468 Supplier Leinco Technologies Inc Supplier No. 12532100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dicer Monoclonal antibody specifically detects Dicer in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen Dicer
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone S167-7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene DICER1
Gene Accession No. Q8R418, Q9UPY3
Gene Alias 1110006F08Rik; D12Ertd7e; DCR; DCR1; DCR-1; Dicer; dicer 1 ribonuclease III; dicer 1, double-stranded RNA-specific endoribonuclease; dicer 1, ribonuclease III; dicer 1, ribonuclease type III; Dicer1; Dicer1, Dcr-1 homolog; Dicer1e; double-strand-specific ribonuclease mDCR-1; endoribonuclease Dicer; Helicase MOI; helicase with RNAse motif; helicase-moi; HERNA; K12H4.8-LIKE; KIAA0928; mKIAA0928; MNG1; RMSE2
Host Species Mouse
Immunogen Fusion protein corresponding to aa 1638-1899 of mouse Endoribonuclease Dicer (accession no.Q8R418).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192119, 23405, 299284
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.