Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicer Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501561359
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-1359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-1359 Supplier Bethyl Laboratories, Inc Supplier No. A301937A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

DICER1 is a ribonuclease that has been extensively shown to be related to cancer. Small interfering RNAs are generated through DICER, and seem to play important roles in mitosis in normal cells and oncogenesis in mutated cells. DICER1 mutations are associated with blastoma, breast cancer, adenocarcinoma, and pancreatitis. In vision and hearing, DICER1 has been implicated in serious microRNA induced malformations of critical sensory cells. DICER1 is important in myelination and other nervous system protections. Finally, DICER1 is related to cardiovascular diseases, since microRNAs are aberrantly expressed in some pathological conditions.

Specifications

Antigen Dicer
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene DICER1
Gene Accession No. Q9UPY3
Gene Alias DCR1, Dicer, HERNA, KIAA0928
Gene Symbols DICER1
Host Species Rabbit
Immunogen Between 1175 and 1225
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23405
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.