Alfa Aesar™ Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II), 98%

Manufacturer: Alfa Aesar™ 1393204

Quantity 2g

 View more versions of this product

Catalog No. AA1393204

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14647-23-5
    Formula Weight 528.02
    MDL Number MFCD00013313
    Synonym 1,2-Bis(diphenylphosphino)ethane nickel(II) chloride
    Chemical Name or Material Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II)

    Specifications

    Formula Weight 528.02
    Health Hazard 2 GHS H Statement
    H351-H302-H312-H332-H315-H319-H317-H335
    Suspected of causing cancer.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    MDL Number MFCD00013313
    Melting Point 263° to 265°C
    Synonym 1,2-Bis(diphenylphosphino)ethane nickel(II) chloride
    Health Hazard 1 Warning
    Color Orange-red to brown
    CAS 14647-23-5
    Quantity 2g
    Chemical Name or Material Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II)
    Assay 98%

    GHS H Statement
    H351-H302-H312-H332-H315-H319-H317-H335
    Suspected of causing cancer.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit