Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II), Pd 17.6%

Catalog No. AA4497103
Click to view available options
Quantity:
1 g
5 g

Catalog No. AA4497103

Supplier: Thermo Scientific Chemicals 04497103

Only null left
Add to Cart

CAS: 29964-62-3 | C28H28Cl2P2Pd | 603.80 g/mol

Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II) is used for the catalysis of styrene carbonylation, coupling of alkyl Grignard reagents with organic halides, selective monoalkylation of organic polyhalides, and modification of the dihalovinyl moiety of synthetic pyrethroids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29964-62-3
Molecular Formula C28H28Cl2P2Pd
Molecular Weight (g/mol) 603.80
MDL Number MFCD02093437
InChI Key JQXJBXVWVPVTOO-UHFFFAOYSA-L
Synonym 1,4-bis diphenylphosphino butane-palladium ii chloride, 1,4-butylenebis diphenylphosphine palladium dichloride, pdcl2 dppb, 1,4-bis diphenylphosphino butane palladium ii dichloride, palladium ii chloride-1,4-bis diphenylphosphine butane, dichloro bis 1,4-diphenylphosphino butane palladium ii, palladium ii chloride 1,4-bis diphenylphosphino butane complex, dichloro 1,4-bis diphenylphosphino butane palladium ii, pd dppb cl2, acmc-209sk1
PubChem CID 10974057
IUPAC Name dichloropalladium;4-diphenylphosphanylbutyl(diphenyl)phosphane
SMILES Cl[Pd]Cl.C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Solubility Information Soluble in ethanol,benzene,and ethereal solvents.
Formula Weight 603.8
Concentration or Composition (by Analyte or Components) Pd 17.6%
Physical Form Lumps
Chemical Name or Material Dichloro[bis (1,4-diphenylphosphino)butane]palladium (II)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.