Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dichloro(chloromethyl)methylsilane, 95%
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Straw |
| Density | 1.2600g/mL |
| Boiling Point | 121.0°C to 122.0°C |
| Flash Point | 35°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4480 to 1.4510 |
| Linear Formula | ClCH2Si(Cl2)CH3 |
| Quantity | 25 g |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Toxic if inhaled. Flammable liquid and vapor. Harmful to aquatic life with long lasting effects. Reacts violently with water. Corrosive to the respiratory tract |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye p |
| EINECSNumber | 216-319-5 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction