Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloro(diethylamino)phosphine, 97%

Catalog No. AAH2593406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2593406 5 g
AAH2593414 25 g
AAH2593422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2593406 Supplier Thermo Scientific Chemicals Supplier No. H2593406
Add to Cart

CAS: 1069-08-5 | C4H10Cl2NP | 174.005 g/mol

Dichloro(diethylamino)phosphine is used as a reagent for the preparation of diethylaminophosphine-metal complexes and it is a critical starting material in the preparation of various phosphines, phosphoramidites & the phosphitylation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichloro(diethylamino)phosphine is used as a reagent for the preparation of diethylaminophosphine-metal complexes and it is a critical starting material in the preparation of various phosphines, phosphoramidites & the phosphitylation of alcohols.

Solubility
Reacts violently in water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, active metals, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1069-08-5
Molecular Formula C4H10Cl2NP
Molecular Weight (g/mol) 174.005
MDL Number MFCD00013614
InChI Key BPEMCEULJQTJMI-UHFFFAOYSA-N
Synonym diethylphosphoramidous dichloride, dichloro diethylamino phosphine, diethylaminodichlorophosphine, et2npcl2, acmc-2098mq, diethylamido dichlorophosphite, diethylamino dichlorophosphine, dichloro diethylamido phosphine, dichlorophosphanyl diethylamine, dichlorophosphorous diethylamide
PubChem CID 2733289
IUPAC Name N-dichlorophosphanyl-N-ethylethanamine
SMILES CCN(CC)P(Cl)Cl

Specifications

Density 1.196
Boiling Point 179°C
Flash Point 83°C (181°F)
Odor Irritating
Refractive Index 1.497
Linear Formula (CH3CH2)2NPCl2
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts violently in water.
Formula Weight 174.01
Percent Purity 97%
Chemical Name or Material Dichloro(diethylamino)phosphine
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.