Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloro(ethylenediamine)palladium(II), Pd 44.8%

Catalog No. p-7048422
Click to view available options
Quantity:
1 g

CAS: 15020-99-2 | C2H8Cl2N2Pd | 237.42 g/mol

Catalyst for C-C coupling reactions like Sonogashira coupling. Formation of host-guest complexes. Synthesis of mixed-metal, mixed-pyrimidine self-assembling metallacalix[n]arenes; ring-in-ring complex; binding of metal with glycose phosphates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15020-99-2
Molecular Formula C2H8Cl2N2Pd
Molecular Weight (g/mol) 237.42
MDL Number MFCD00044991
InChI Key CAYKJANQVKIYPJ-UHFFFAOYSA-L
Synonym dichloro 1,2-diaminoethane palladium, dichloro ethylenediamine palladium ii, dichloro 1,2-diaminoethane ;palladium, dichloro ethylenediamine palladate ii, ethylenediamine palladium ii chloride, ethylenediamine; palladium chloride, palladium ii chloride ethylenediamine complex, dichloro ethylenediamine palladium ii , pd
PubChem CID 11139157
SMILES Cl[Pd]Cl.NCCN

Specifications

Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Very low solubility in water.
IUPAC Name dichloropalladium; ethane-1,2-diamine
Formula Weight 237.41
Concentration or Composition (by Analyte or Components) Pd 44.8%
Physical Form Crystalline
Chemical Name or Material Dichloro(ethylenediamine)palladium(II)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.