Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer, Thermo Scientific™

Catalog No. AC363320010
Click to view available options
:
1g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 96503-27-4
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C20H30Cl4Ru2
Quantity 1g
MDL Number MFCD00070449
Synonym dichloro pentamethylcyclopentadienyl ruthenium iii polymer, pentamethylcyclopentadienylruthenium iii chloride polymer, dichloro pentamethylcyclopentadienyl ruthenium iii , polymer
InChI Key VDXZXHGAMPUNED-UHFFFAOYSA-J
SMILES C[C]1[C]([C]([C]([C]1C)C)C)C.C[C]1[C]([C]([C]([C]1C)C)C)C.Cl[Ru]Cl.Cl[Ru]Cl
IUPAC Name dichlororuthenium;1,2,3,4,5-pentamethylcyclopentane
Molecular Weight (g/mol) 614.4
PubChem CID 72376328
Chemical Name or Material Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.