Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloroacetic acid, 99+%

Catalog No. p-3736169
Click to view available options
Quantity:
2.5 L
Packaging:
Glass bottle

CAS: 79-43-6 | C2H2Cl2O2 | 128.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-43-6
Molecular Formula C2H2Cl2O2
Molecular Weight (g/mol) 128.94
MDL Number MFCD00004223
InChI Key JXTHNDFMNIQAHM-UHFFFAOYSA-N
Synonym dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid
PubChem CID 6597
ChEBI CHEBI:36386
IUPAC Name 2,2-dichloroacetic acid
SMILES C(C(=O)O)(Cl)Cl

Specifications

Melting Point 9.0°C to 11.0°C
Color Brown
Density 1.5600g/mL
Boiling Point 194.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4653 to 1.4673
Linear Formula Cl2CHCO2H
Quantity 2.5 L
Beilstein 02,202
Fieser 16,115
Merck Index 15,3059
Specific Gravity 1.56
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol,ether and ethanol,soluble in acetone
Formula Weight 128.94
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Dichloroacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Suspected of causing cancer.
May damage fertility or the unborn child.
May cause harm to breast-fed children.
May cause damage to or
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 201-207-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.