Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloroacetonitrile, 98+%

Catalog No. AAA1061222
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 3018-12-0 | C2HCl2N | 109.937 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3018-12-0
Molecular Formula C2HCl2N
Molecular Weight (g/mol) 109.937
MDL Number MFCD00040886
InChI Key STZZWJCGRKXEFF-UHFFFAOYSA-N
Synonym dichloroacetonitrile, acetonitrile, dichloro, dichloromethyl cyanide, ccris 942, unii-o0l64v63m9, dichloro acetonitrile, pubchem17242, acmc-209hdn, dsstox_cid_1562
PubChem CID 18177
ChEBI CHEBI:82444
IUPAC Name 2,2-dichloroacetonitrile
SMILES C(#N)C(Cl)Cl

Specifications

Density 1.369
Boiling Point 112°C to 114°C
Flash Point 35°C (95°F)
Odor Pungent
Refractive Index 1.4405
Linear Formula Cl2CHCN
Quantity 100 g
UN Number UN2920
Beilstein 1739029
Formula Weight 109.94
Percent Purity ≥98%
Chemical Name or Material Dichloroacetonitrile
Show More Show Less
Hazard Category H226-H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226-H341-H361-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 221-159-4
RTECSNumber AL8465000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.