Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloroacetyl chloride, 97%

Catalog No. p-7026341
Click to view available options
Quantity:
100 g
500 g

CAS: 79-36-7 | C2HCl3O | 147.379 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-36-7
Molecular Formula C2HCl3O
Molecular Weight (g/mol) 147.379
MDL Number MFCD00000840
InChI Key FBCCMZVIWNDFMO-UHFFFAOYSA-N
Synonym dichloroacetyl chloride, dichloracetyl chloride, acetyl chloride, dichloro, dichloroethanoyl chloride, chlorure de dichloracetyle, dichloroacetylchloride, dichloroacetic acid chloride, alpha,alpha-dichloroacetyl chloride, chcl2cocl, chlorid kyseliny dichloroctove
PubChem CID 6593
ChEBI CHEBI:34688
IUPAC Name 2,2-dichloroacetyl chloride
SMILES C(C(=O)Cl)(Cl)Cl

Specifications

Density 1.532
Boiling Point 107°C to 108°C
Flash Point None
Refractive Index 1.462
Quantity 100 g
UN Number UN1765
Sensitivity Moisture sensitive
Merck Index 14,3053
Formula Weight 147.39
Percent Purity 97%
Chemical Name or Material Dichloroacetyl chloride
Show More Show Less
Hazard Category H227-H311-H314-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DICHLOROACETYL CHLORIDE
EINECSNumber 201-199-9
RTECSNumber AO6650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.