Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorodiethylsilane, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1719-53-5
C4H10Cl2Si
157.12
MFCD00000493
BYLOHCRAPOSXLY-UHFFFAOYSA-N
diethyldichlorosilane,silane, dichlorodiethyl,dichloro diethyl silane,diethyldichlorosilicon,diethyl dichlorosilane,diethyidichlorosilane,snpladqptpdumup@,dichlorodiethyl-silan,diethylsiliciumdichlorid,silane,dichlorodiethyl
15589
dichloro(diethyl)silane
CC[Si](CC)(Cl)Cl
1719-53-5
C4H10Cl2Si
157.12
MFCD00000493
BYLOHCRAPOSXLY-UHFFFAOYSA-N
diethyldichlorosilane,silane, dichlorodiethyl,dichloro diethyl silane,diethyldichlorosilicon,diethyl dichlorosilane,diethyidichlorosilane,snpladqptpdumup@,dichlorodiethyl-silan,diethylsiliciumdichlorid,silane,dichlorodiethyl
15589
dichloro(diethyl)silane
CC[Si](CC)(Cl)Cl
Flash Point 28°C
Specific Gravity 1.05
Packaging Glass bottle
Density 1.0500g/mL
Quantity 5g
Boiling Point 128.0°C to 129.0°C
Melting Point -96.0°C
Assay Percent Range 97%
Linear Formula (CH3CH2)2Si(Cl)2
Solubility Information Solubility in water: reacts
Formula Weight 157.12
Percent Purity 97%
Chemical Name or Material Dichlorodiethylsilane, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 217-005-0

RTECSNumber : VV3060000

TSCA : TSCA