Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorofluorescein, TS, Reagents

Catalog No. 01335428
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-428 120 mL
01-335-427 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-428 Supplier Reagents Holdings Llc Supplier No. CD226600120A
Only null left
Add to Cart
Add to Cart

Dichlorofluorescein, TS, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 7732-18-5 , 67-63-0
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym 2',7'-Dichloro-3,6-fluorandiol, 2,7-Dichlorofluorescein, 2',7'-dichloro-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
IUPAC Name ethanol
SMILES CCO

Specifications

Boiling Point 82°C
Grade USP
Packaging Poly Bottle
Specific Gravity 0.9
Color Yellow/Green
Quantity 120 mL
Physical Form Liquid
Chemical Name or Material Dichlorofluorescein, TS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.