Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
500mL
1L
2.5L
Packaging:
AcroSeal glass Bottle
AcroSeal™ Glass Bottle
Glass bottle
75-09-2,513-35-9
CH2Cl2
84.93
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
75-09-2,513-35-9
CH2Cl2
84.93
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride,methylene dichloride,methane, dichloro,methylene bichloride,methane dichloride,solaesthin,solmethine,freon 30,narkotil,aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
Specific Gravity 1.325
Viscosity 0.43 mPa.s (20°C)
Melting Point -97.0°C
Water 0.005% Max. (Coulometric)
Vapor Pressure 475hPa at 20°C
Stabilizer 50ppm (approx.) amylene
Quantity 100mL
Infrared Spectrum Authentic
Decomposition Information >120°C
Density 1.3250g/mL
Color Colorless
Packaging AcroSeal™ Glass Bottle
Boiling Point 39.0°C to 40.0°C
Assay Percent Range 99.8% min. (GC)
Linear Formula CH2Cl2
Beilstein 01, 60
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Formula Weight 84.93
Physical Form Liquid
Percent Purity 99.8%
Grade Extra Dry over Molecular Sieve
Chemical Name or Material Dichloromethane
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer.
May cause drowsiness or dizziness.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective cloth

GHS Signal Word: Warning

EINECSNumber : 200-838-9