Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, 99.8%, for HPLC, stabilized with methanol

Catalog No. p-217032
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle

Catalog No. AC364230010

Supplier: Thermo Scientific Chemicals 364230010

Only null left
Add to Cart

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2, 67-56-1
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97.0°C
Absorbance (1 cm cell vs HPLC−grade water), 0.005 max. at 260nm, 0.005 max. at 270nm, 0.005 max. at 280nm, 0.02 max. at 250nm, 0.2 max. at 240nm, 1 max. at 233nm
Acidity 0.0001 meq/g max.
Density 1.3250g/mL
Boiling Point 39.0°C to 40.0°C
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.0002% max.
Assay Percent Range 99.80%
Specific Gravity 1.325
Linear Formula CH2Cl2
Viscosity 0.43 mPa.s (20°C)
Beilstein 01, 60
Water 0.02% max.
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Formula Weight 84.93
Percent Purity 99.8%
Grade HPLC
Chemical Name or Material Dichloromethane, Stabilized
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Obtain special instructions before use. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-838-9
RTECSNumber PA8050000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.