Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, 99.9%, Extra Dry, AcroSeal™, 99.9%
GSA_VA

Catalog No. AC610300010
Click to view available options
:
1L
This item is not returnable. View return policy

Catalog No. AC610300010

Supplier: Thermo Scientific Chemicals 610300010

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

CAS 75-09-2, 513-35-9
Decomposition Information >120°C
Density 1.3250g/mL
Quantity 1 L
Identification Pass Test
Assay Percent Range 99.9%
Molecular Formula CH2Cl2
Linear Formula CH2Cl2
Vapor Pressure Vapor Pressure: 475hPa at 20°C
MDL Number MFCD00000881
Beilstein 01, 60
Water 50ppm max.
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES ClCCl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
PubChem CID 6344
ChEBI CHEBI:15767
Formula Weight 84.93
Percent Purity ≥99.0%
Grade Extra Dry
Chemical Name or Material Dichloromethane, Anhydrous
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-838-9
RTECSNumber PA8050000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.