Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, 99+%, extra pure, stabilized with ethanol

Catalog No. AC124050050
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2, 64-17-5
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Density 1.3250g/mL
Quantity 5 L
Packaging Plastic Bottle
Physical Form Liquid
Residue after Evaporation 0.05% max.
Assay Percent Range 99+%
Linear Formula CH2Cl2
Beilstein 01, 60
Water 0.1% max. (K.F.)
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Formula Weight 84.93
Percent Purity ≥99.0%
Grade Extra Pure
Chemical Name or Material Dichloromethane
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Use personal protective equipment as required. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.