Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dichloromethane ChromAR, Macron Fine Chemicals™
Shop All Avantor Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass
SAFEMOR
Specifications
Specifications
| Quantity | 1 L |
| Packaging | Glass |
| Formula Weight | 84.93 |
| Chemical Name or Material | Dichloromethane |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction