Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dichloromethane, For pesticide residue analysis, Honeywell Riedel-de Haën™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent

Specifications
Specifications
Melting Point | -97°C (literature) |
CAS Min % | ≥99.0000% |
CAS Max % | ≥99.0000% |
Density | 1.325 g/mL at 25 °C(lit.) |
Boiling Point | 39°C to 41°C |
Quantity | 4 x 4 L |
Packaging | Glass Bottle |
Linear Formula | CH2Cl2 |
UN Number | UN1593 |
Beilstein | 1730800 |
Show More |
Safety and Handling
ShelfLife | 720 days from date of manufacture |
DOTInformation | Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Dichloromethane |
EINECSNumber | 200-838-9 |
Recommended Storage | Room Temp |
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction