Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane-d2, for NMR, 99.5 atom % D

Catalog No. AC176110250
Click to view available options
Quantity:
5 mL
10 mL
25 mL
50 mL
Packaging:
Glass Bottle

CAS: 1665-00-5 | CH2Cl2 | 86.939 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1665-00-5
Molecular Formula CH2Cl2
MDL Number MFCD00000882
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
Synonym dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
PubChem CID 160586

Specifications

Physical Form Liquid
Refractive Index 1.4218
Color Colorless
Packaging Glass Bottle
Quantity 25 mL
Density 1.3600g/mL
Water 0.04% max. (Coulometric)
Deuterium (D) 99.60% min.
Melting Point -97.0°C
Boiling Point 40.0°C
Linear Formula CD2Cl2
Beilstein 01, IV, 39
SMILES C(Cl)Cl
IUPAC Name dichloro(dideuterio)methane
Molecular Weight (g/mol) 86.939
Formula Weight 86.95
Grade NMR
Chemical Name or Material Dichloromethane-d2, 99.6+ atom % D
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-776-0

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.