Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane For spectroscopy Uvasol™, MilliporeSigma™

Catalog No. m1060480500
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle

For spectroscopy Uvasol™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

pH Neutral (at 20°C)
Melting Point -95°C
Color Colorless
Density 1.33g/cm3 at 20°C
Boiling Point 40°C
Quantity 500 mL
Ignition Point 605°C (1121°F)
Odor Sweet
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.42
Vapor Pressure 475 hPa (20°C)
UN Number 1593
Solubility Information Soluble in water: 20g/L at 20°C
Formula Weight 84.93
Chemical Name or Material Dichloromethane
Show More Show Less
Hazard Category H351: Suspected of causing cancer.
Recommended Storage 5°C to 30°C (41°F to 86°F), Tightly closed. Keep in a well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized persons. Protected from light.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.