Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethylphosphine, 90%, Tech., Thermo Scientific Chemicals

Catalog No. p-3778020
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 676-83-5
Molecular Formula CH3Cl2P
Molecular Weight (g/mol) 116.909
MDL Number MFCD00013616
InChI Key CDPKWOKGVUHZFR-UHFFFAOYSA-N
Synonym methyldichlorophosphine, dichloromethylphosphine, methylphosphonous dichloride, dichloro methyl phosphane, phosphonous dichloride, p-methyl, ch3pcl2, phosphonous dichloride, methyl, methyl phosphonous dichloride, methylphosphorus dichloride, phosphine, dichloromethyl
PubChem CID 61194
IUPAC Name dichloro(methyl)phosphane
SMILES CP(Cl)Cl

Specifications

Density 1.3000g/mL
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula CH3PCl2
Quantity 10 g
Formula Weight 116.92
Grade Technical
Chemical Name or Material Dichloromethylphosphine
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed. Harmful if inhaled. Causes severe skin burns and eye damage. Catches fire spontaneously if exposed to air. Reacts violently with water. Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not allow contact with air.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.