Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethylsilane, 97+%

Catalog No. p-7049345
Click to view available options
Quantity:
100 g
500 g

CAS: 75-54-7

A monomer (‘building block’) in the production of silicone polymers. An intermediate (starting material) in the production of other organic and inorganic chemicals. In the electronics industry for the production of ultra-pure polysilicon and in the manufacture of semiconductors and photovoltaics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-54-7
MDL Number MFCD00000494
Synonym Methyldichlorosilane

Specifications

Boiling Point 41°C to 42°C
Melting Point -93°C
Color Colorless
Physical Form Liquid
Quantity 100 g
Linear Formula CH3SiHCl2
UN Number UN1242
Beilstein 1071194
Flash Point -32°C (-26°F)
Solubility Information Soluble in heptane,benzene,ether.
Formula Weight 115.03
Percent Purity ≥97%
Refractive Index 1.398
Sensitivity Air and moisture sensitive
Density 1.105 g/mL
Chemical Name or Material Dichloromethylsilane
Show More Show Less
Hazard Category H225-H260-H301-H314-H330
Hazard Statement GHS H Statement
H225-H260-H314-H318-H332
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P210-P223-P231+P232-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: METHYLDICHLOROSILANE
EINECSNumber 200-877-1
RTECSNumber VV3500000
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.