Alfa Aesar™ Dichlorophenylphosphine, 97%

Manufacturer: Alfa Aesar™ A1028430

Quantity 250g

 View more versions of this product

Catalog No. AAA1028430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 644-97-3
    Molecular Formula C6H5Cl2P
    Formula Weight 178.99
    MDL Number MFCD00000528
    UN Number UN2798
    Beilstein 508189
    Synonym Phenylphosphine dichloride; Phenylphosphonous dichloride
    Chemical Name or Material Dichlorophenylphosphine

    Specifications

    Formula Weight 178.99
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C6H5Cl2P
    CAS 644-97-3
    Health Hazard 2 GHS H Statement
    H300-H314-H318
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    TSCA TSCA
    MDL Number MFCD00000528
    UN Number UN2798
    Flash Point 102°C (215°F)
    Synonym Phenylphosphine dichloride; Phenylphosphonous dichloride
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHENYL PHOSPHORUS DICHLORIDE
    RTECS Number TB2478000
    Quantity 250g
    Odor Pungent
    Boiling Point 99° to 101°C (11mmHg)
    Beilstein 508189
    Melting Point −51°C
    Chemical Name or Material Dichlorophenylphosphine
    Assay 97%

    Description

    Applications
    Dichlorophenylphosphine and its derivatives are used as an intermediate to make plasticizers, antioxidants, stabilizers, catalysts and high pressure lubricant additives and in lacquer formulatings.

    Solubility
    Reacts with water.

    Notes
    Air & moisture Sensitive. Incompatible with bases, oxidizing agents, water/ moisture &air. Store under inert gas.

    GHS H Statement
    H300-H314-H318
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit