Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorophenylsilane, 96%

Catalog No. p-7044071
Click to view available options
Quantity:
5 g
25 g

CAS: 1631-84-1 | C6H5Cl2Si | 176.09 g/mol

Dichlorophenylsilane is used as the intermediates of OLED,pharmaceuticals, cosmetics. It is also used as reduction reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichlorophenylsilane is used as the intermediates of OLED,pharmaceuticals, cosmetics. It is also used as reduction reagent.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from moisture, oxidizing agents. Incompatible materials are bases, moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1631-84-1
Molecular Formula C6H5Cl2Si
Molecular Weight (g/mol) 176.09
MDL Number MFCD00039296
InChI Key XNAFLNBULDHNJS-UHFFFAOYSA-N
Synonym phenyldichlorosilane, dichlorophenylsilane, dichloro phenyl silane, dichloro phenyl silicon, benzene, dichlorosilyl, phenyl dichlorosilane, phenyl hydrogen dichlorosilane, phenyl-dichlorosilane, phenyldichloro silane, dichloro phenyl silyl
PubChem CID 6327324
SMILES Cl[Si](Cl)C1=CC=CC=C1

Specifications

Boiling Point 181°C
Quantity 5 g
UN Number UN2986
Beilstein 2433865
Flash Point 48°C (118°F)
Solubility Information Hydrolyzes in water.
IUPAC Name dichloro(phenyl)silicon
Formula Weight 177.11
Percent Purity 96%
Refractive Index 1.523
Sensitivity Moisture sensitive
Density 1.212
Chemical Name or Material Dichlorophenylsilane
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 216-635-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.