Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorophenylsilane, 95%

Catalog No. ac432340250
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle

CAS: 1631-84-1 | C6H5Cl2Si | 176.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1631-84-1
Molecular Formula C6H5Cl2Si
Molecular Weight (g/mol) 176.09
MDL Number MFCD00039296
InChI Key XNAFLNBULDHNJS-UHFFFAOYSA-N
Synonym phenyldichlorosilane, dichlorophenylsilane, dichloro phenyl silane, dichloro phenyl silicon, benzene, dichlorosilyl, phenyl dichlorosilane, phenyl hydrogen dichlorosilane, phenyl-dichlorosilane, phenyldichloro silane, dichloro phenyl silyl
PubChem CID 6327324
IUPAC Name dichloro(phenyl)silyl
SMILES Cl[Si](Cl)C1=CC=CC=C1

Specifications

Color White to Yellow
Density 1.2040g/mL
Boiling Point 65.0°C to 66.0°C (10.0 mmHg)
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5245 to 1.5275 (20°C, 589nm)
Quantity 25 mL
Specific Gravity 1.204
Formula Weight 177.11
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Dichlorophenylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 216-635-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.