Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

μ-Dichlorotetraethylene dirhodium(I)

Catalog No. AC197590010
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle

CAS: 12081-16-2 | C8H16Cl2Rh2 | 388.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12081-16-2
Molecular Formula C8H16Cl2Rh2
Molecular Weight (g/mol) 388.93
MDL Number MFCD00013206
InChI Key OOQJCPOXJFCGCR-UHFFFAOYSA-L
Synonym chlorobis ethylene rhodium i dimer, dichlorotetra ethylene dirhodium i
PubChem CID 45357812
IUPAC Name ethene;rhodium;dichloride
SMILES Cl[Rh].Cl[Rh].C=C.C=C.C=C.C=C

Specifications

Color Rust
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 388.92
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material µ-Dichlorotetraethylene dirhodium(I)
Hazard Category AIR SENSITIVE
EINECSNumber 235-145-0

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.