Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorotris(triphenylphosphine)ruthenium(II), 97%

Catalog No. AAL0037306
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 15529-49-4 | C54H45Cl2P3Ru | 958.85 g/mol

Dichlorotris(triphenylphosphine)ruthenium(II) is used as a catalyst employed in a synthesis of furans from allenyl sulfides. C-H Activation Catalyst in a Ruthenium/Lewis Acid System. It is used in the Kharasch addition of chlorocarbons to alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichlorotris(triphenylphosphine)ruthenium(II) is used as a catalyst employed in a synthesis of furans from allenyl sulfides. C-H Activation Catalyst in a Ruthenium/Lewis Acid System. It is used in the Kharasch addition of chlorocarbons to alkenes.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15529-49-4
Molecular Formula C54H45Cl2P3Ru
Molecular Weight (g/mol) 958.85
MDL Number MFCD00013077
InChI Key KSHWMFNJCNGTJO-UHFFFAOYSA-L
Synonym dichlorotris triphenylphosphine ruthenium ii, tris triphenylphosphine ruthenium ii chloride, dichlorotris triphenylphosphino ruthenium ii, tris triphenylphosphine ruthenium ii dichloride, dichlorotris triphenylphosphine ruthenium, ruthenium ii-tris triphenylphosphine dichloride, tris triphenylphosphine dichlororuthenium, ruthenium dichlorotris triphenylphosphine, pubchem12822
PubChem CID 11007548
SMILES [Cl-].[Cl-].[Ru+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 132°C to 134°C
Quantity 5 g
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
IUPAC Name dichlororuthenium;triphenylphosphane
Formula Weight 958.86
Percent Purity 97%
Chemical Name or Material Dichlorotris(triphenylphosphine)ruthenium(II)
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 239-569-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.