Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dickson™ AC Adapter for Data Loggers
SDP

Compatible with all Dickson units

Supplier:  Dickson™ R157

Catalog No. 15-075-249


Description

Description

  • Input: 120 to 240V
  • Output: 12VDC
  • Weight: 2 lb.

Specifications

Specifications

Adapter, AC
Supports easy data collection
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.