Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Diclofenac Monoclonal Antibody (3B6)
GREENER_CHOICE

Catalog No. PIMA549626
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549626 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549626 Supplier Invitrogen™ Supplier No. MA549626
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Diclofenac is a gene target that is associated with the metabolism of the nonsteroidal anti-inflammatory drug (NSAID) diclofenac. This gene is involved in the breakdown of diclofenac in the liver, which can affect the drug's efficacy and toxicity. Diclofenac is commonly used to treat pain and inflammation, but it can also have harmful side effects on the liver and gastrointestinal system. Understanding the role of Diclofenac in diclofenac metabolism can help researchers develop safer and more effective NSAIDs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Diclofenac
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3B6
Concentration 12.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of DC hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.