Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicobalt octacarbonyl, 95%, stabilized

Catalog No. p-107363
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 10210-68-1 | C8Co2O8 | 341.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10210-68-1
Molecular Formula C8Co2O8
Molecular Weight (g/mol) 341.95
MDL Number MFCD00016024
Synonym carbon monoxide; cobalt 2+, dicobalt octacarbonyl stabilised with hexane, cobalt carbonyl, moistened with hexane hexane co

Specifications

Melting Point 51.0°C to 52.0°C
Color Orange-Red to Red
Density 1.8100g/mL
Flash Point −13°C
Infrared Spectrum Authentic
Assay Percent Range 94% min
Packaging Glass bottle
Linear Formula Co2(CO)8
Quantity 25 g
Merck Index 15,3095
Specific Gravity 1.81
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and carbon disulfide
Stabilizer 5% max. hexane
Formula Weight 341.95
Percent Purity 95%
Physical Form Crystals or Flakes
Chemical Name or Material Dicobalt octacarbonyl, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Harmful if inhaled.
Suspected of causing cancer.
Harmful to aquatic life with long lasting effects.
Self-heating; may catch fire.
Precautionary Statement GHS P Statement
Keep cool.
Protect from sunlight.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty
EINECSNumber 233-514-0
RTECSNumber GG0300000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.