Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dicumyl peroxide, 99%, ACROS Organics™
Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
Description
Radical initiators
Specifications
Infrared Spectrum | Authentic |
Flash Point | >110°C |
Melting Point | 38.0°C to 41.0°C |
Quantity | 100g |
Packaging | Plastic bottle |
Color | White |
Assay Percent Range | 98.5% min. (Iodometry) |
Linear Formula | [C6H5C(CH3)2]2O2 |
Solubility Information | Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents |
Formula Weight | 270.37 |
Show More |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Heating may cause a fire.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face p
GHS Signal Word: Warning
EINECSNumber : 201-279-3
RTECSNumber : SD8150000
TSCA : TSCA