Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dicyanine A 98%, Thermo Scientific™
Click to view available options
:
1g; Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
CAS | 20591-23-5 |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Molecular Formula | C25H25IN2 |
Quantity | 1g |
MDL Number | MFCD00051379 |
Beilstein | 23, IV, 2069 |
Synonym | dicyanine a |
InChI Key | AIUPVZRNWAATQJ-UHFFFAOYSA-M |
Show More |
Safety and Handling
EINECSNumber | 243-897-6 |
TSCA | TSCA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction