Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium(III) chloride, anhydrous, 48.6-49.2% Rh

Catalog No. AC419100025
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle

CAS: 10049-07-7 | Cl3Rh | 209.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-07-7
Molecular Formula Cl3Rh
Molecular Weight (g/mol) 209.26
MDL Number MFCD00011204
InChI Key SONJTKJMTWTJCT-UHFFFAOYSA-K
IUPAC Name rhodium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Rh+3]

Specifications

Boiling Point 717.0°C (730.0 mmHg)
Melting Point 450.0°C
Color Black
Physical Form Crystalline Powder
Quantity 2.5 g
Linear Formula RhCl3
Fieser 02,357; 03,242; 06,504; 07,313; 08,435; 11,460; 14,271; 16,292; 17,366
Merck Index 15, 8313
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkaline solution
Formula Weight 209.26
Percent Purity 48.6 to 49.2% Rh
Packaging Glass bottle
Specific Gravity 5.38
Density 5.3800g/mL
Chemical Name or Material Rhodium(III) chloride, 48.6-49.2% Rh
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Suspected of causing genetic defects.
May be corrosive to metals.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 233-165-4
RTECSNumber VI9275000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.