Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicyclohexyl phthalate, 99+%

Catalog No. p-7037296
Click to view available options
Quantity:
100 g
500 g

CAS: 84-61-7 | C20H26O4 | 330.424 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-61-7
Molecular Formula C20H26O4
Molecular Weight (g/mol) 330.424
MDL Number MFCD00003849
InChI Key VOWAEIGWURALJQ-UHFFFAOYSA-N
Synonym dicyclohexyl phthalate, ergoplast fdc, unimoll 66, ergoplast.fdc, howflex cp, phthalic acid, dicyclohexyl ester, phthalic acid dicyclohexyl ester, 1,2-benzenedicarboxylic acid, dicyclohexyl ester, unii-cgd15m7h2n, ccris 6190
PubChem CID 6777
ChEBI CHEBI:34693
IUPAC Name dicyclohexyl benzene-1,2-dicarboxylate
SMILES C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3

Specifications

Melting Point 63°C to 67°C
Density 1.383
Refractive Index 1.431
Quantity 100 g
Beilstein 1889288
Formula Weight 330.42
Percent Purity ≥99%
Chemical Name or Material Dicyclohexyl phthalate
Hazard Category H317-H360D
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 201-545-9
RTECSNumber TI0889000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.