Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicyclohexyl phthalate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
250g
1kg
2.5kg
Packaging:
Glass bottle
84-61-7
C20H26O4
330.42
MFCD00003849
VOWAEIGWURALJQ-UHFFFAOYSA-N
dicyclohexyl phthalate,ergoplast fdc,unimoll 66,ergoplast.fdc,howflex cp,phthalic acid, dicyclohexyl ester,phthalic acid dicyclohexyl ester,1,2-benzenedicarboxylic acid, dicyclohexyl ester,unii-cgd15m7h2n,ccris 6190
6777
CHEBI:34693
dicyclohexyl benzene-1,2-dicarboxylate
C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3
84-61-7
C20H26O4
330.42
MFCD00003849
VOWAEIGWURALJQ-UHFFFAOYSA-N
dicyclohexyl phthalate,ergoplast fdc,unimoll 66,ergoplast.fdc,howflex cp,phthalic acid, dicyclohexyl ester,phthalic acid dicyclohexyl ester,1,2-benzenedicarboxylic acid, dicyclohexyl ester,unii-cgd15m7h2n,ccris 6190
6777
CHEBI:34693
dicyclohexyl benzene-1,2-dicarboxylate
C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3
Quantity 5g
Color White
Flash Point 207°C
Boiling Point 200.0°C to 235.0°C (4.0mmHg)
Melting Point 63.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Density 207g/mL
Assay Percent Range 98.5% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 330.42
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Dicyclohexyl phthalate
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
May damage the unborn child.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention

GHS Signal Word: Danger

EINECSNumber : 201-545-9