Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicyclohexylamine Borane 95.0+%, TCI America™
SDP

Catalog No. B554525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B554525G 25 g
B55455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B554525G Supplier TCI America Supplier No. B554525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 131765-96-3
Molecular Formula C12H26BN
Synonym Borane - Dicyclohexylamine Complex

Specifications

Melting Point 118°C
Color White
Quantity 25 g
Formula Weight 195.16
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dicyclohexylamine Borane
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.