Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Didecylmethylamine 95.0+%, TCI America™
SDP

Catalog No. D2311500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2311500ML 500 mL
D231125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2311500ML Supplier TCI America Supplier No. D2311500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7396-58-9
Molecular Formula C21H45N
Molecular Weight (g/mol) 311.598
MDL Number MFCD00077727
InChI Key ATBNMWWDBWBAHM-UHFFFAOYSA-N
PubChem CID 81880
IUPAC Name N-decyl-N-methyldecan-1-amine
SMILES CCCCCCCCCCN(C)CCCCCCCCCC

Specifications

Melting Point -7°C
Color Yellow
Boiling Point 145°C
Quantity 500 mL
UN Number 2735
Formula Weight 311.60
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Didecylmethylamine
EINECSNumber (2)-0176
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.