Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Didodecylamine 97.0+%, TCI America™
SDP

Catalog No. D026725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D026725G 25 g
D02675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D026725G Supplier TCI America Supplier No. D026725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3007-31-6
Molecular Formula C24H51N
Molecular Weight (g/mol) 353.679
MDL Number MFCD00041918
InChI Key MJCJUDJQDGGKOX-UHFFFAOYSA-N
Synonym didodecylamine, di-n-dodecylamine, 1-dodecanamine, n-dodecyl, dilaurylamine, alamine 204, n-dodecyl-1-dodecanamine, unii-5kr2ml6h0c, 5kr2ml6h0c, dilauryl amine, didodecyl amine
PubChem CID 18157
IUPAC Name N-dodecyldodecan-1-amine
SMILES CCCCCCCCCCCCNCCCCCCCCCCCC

Specifications

Melting Point 51°C
Color White-Yellow
Boiling Point 265°C
Quantity 25 g
Formula Weight 353.68
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Didodecylamine
EINECSNumber (2)-0176&(2)-0138
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.