Learn More
Stoelting™ Dieffenbach Serrefine
SDP
Catalog No. p-7163544 Shop All Stoelting Co Products
Click to view available options
Model:
Curved
Straight
Length (English):
1.2 in.
1.25 in.
Stoelting™ Dieffenbach Serrefine is a useful instrument for stopping blood flow to a particular organ of interest.
250 Grams of pressure
Specifications
For Use With (Application) Neuroscience
Length (English) 1.2 in.
Model Straight
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.