Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethanolamine, 99%

Catalog No. p-7021162
Click to view available options
Quantity:
250 g
500 g
2500 g

CAS: 111-42-2 | C4H11NO2 | 105.14 g/mol

Diethanolamine is used in the preparation of morpholine and diethanolamides, which is an active ingredient in cosmetics and shampoos. It acts as a surfactant and a corrosion inhibitor. It is utilized to remove hydrogen sulfide and carbon dioxide from natural gas. Further, it is an intermediate used in the rubber chemicals, as a humectant and a softening agent, and as an emulsifier and dispersing agent in agricultural chemicals. In addition to this, it is used in cutting oils, cleaners, soaps, polishers, and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethanolamine is used in the preparation of morpholine and diethanolamides, which is an active ingredient in cosmetics and shampoos. It acts as a surfactant and a corrosion inhibitor. It is utilized to remove hydrogen sulfide and carbon dioxide from natural gas. Further, it is an intermediate used in the rubber chemicals, as a humectant and a softening agent, and as an emulsifier and dispersing agent in agricultural chemicals. In addition to this, it is used in cutting oils, cleaners, soaps, polishers, and pharmaceuticals.

Solubility
Miscible with water.

Notes
Hygroscopic and air sensitive. Incompatible with strong oxidizing agents, strong acids, copper, zinc, carbon dioxide and iron.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-42-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
Synonym diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis
PubChem CID 8113
ChEBI CHEBI:28123
IUPAC Name 2-(2-hydroxyethylamino)ethanol
SMILES OCCNCCO

Specifications

Melting Point 27°C to 30°C
Density 1.097
pH 11
Boiling Point 268°C to 270°C
Flash Point 176°C (349°F)
Odor Ammonia-like
Assay Percent Range 99%
Refractive Index 1.477
Quantity 250 g
Beilstein 605315
Sensitivity Hygroscopic
Merck Index 14,3107
Solubility Information Miscible with water.
Formula Weight 105.14
Percent Purity 99%
Chemical Name or Material Diethanolamine
Show More Show Less
Hazard Category H302-H315-H318-H335-H351-H373
Hazard Statement GHS H Statement
H318-H373-H302-H315
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement P201-P202-P234-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P362-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 203-868-0
RTECSNumber KL2975000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.