Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethanolamine, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187392
Click to view available options
Quantity:
5 mL
Diethanolamine (DEA) is an organic chemical compound, which comprises of a secondary amine and a dialcohol. It mainly acts as a weak base. DEA can be used in preparing long-chain fatty acids, namely diethanolamides and diethanolamine salts, which find application in liquid laundry and dishwashing detergents, cosmetics, shampoos, hair conditioners, textile processing, and also in pharmaceutical industries. Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 111-42-2
Ignition Point 689°F
Vapor Pressure <0.98 atm (100°C)
Vapor Density 3.6 (vs. air)
Density 1.097 g/mL (at 25°C)
Flash Point 138°C
Physical Form Neat
Linear Formula HN(CH2 CH2 OH)2
MDL Number MFCD00002843
Melting Point 28°C
Boiling Point 217°C (150 mmHg)
Synonym 2,2′-Iminodiethanol; Bis(2-hydroxyethyl)amine
Type Analytical Standard
Quantity 5 mL
Formula Weight 105.14
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.