Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethanolamine, Reagent, ACS, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18602186
Click to view available options
Quantity:
100 mL
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can

D1033, 111-42-2, C4H11NO2

Spectrum™ Chemical Diethanolamine, Reagent, ACS is also referred to as DEA, is an organic compound that in its pure form is a white solid but it is most often encountered as a clear liquid that has a viscous texture. It is a secondary amine and diol and is water soluble. It is used as a corrosion inhibitor and surfactant that is used in the oil refining industry to help remove hydrogen sulfide from sour gas. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 111-42-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
IUPAC Name 2-[(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCNCCO

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 4 L
Percent Purity 98.5%
Grade ACS
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.